missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SGPP1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£185.00 - £383.00
Specifications
| Antigen | SGPP1 |
|---|---|
| Dilution | Western Blot 1:200 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18313793
|
Novus Biologicals
NBP3-11905-25UG |
25 μg |
£185.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18342783
|
Novus Biologicals
NBP3-11905-100UG |
100 μg |
£383.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SGPP1 Polyclonal antibody specifically detects SGPP1 in Human samples. It is validated for Western BlotSpecifications
| SGPP1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| sphingosine-1-phosphate phosphatase 1 | |
| Synthetic peptide derived from the SGPP1 protein. (Uniprot: Q9BX95) | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot 1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS | |
| 81537 | |
| IgG | |
| Ammonium sulfate precipitation |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title