missing translation for 'onlineSavingsMsg'
Learn More

SGPP1 Antibody, Novus Biologicals™

Product Code. 18346370 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μL
25 μL
Unit Size:
100µL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18346370 100 μL 100µL
18642980 25 μL 25µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18346370 Supplier Novus Biologicals Supplier No. NBP276561

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SGPP1 Polyclonal specifically detects SGPP1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SGPP1
Applications Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunocytochemistry/Immunofluorescence 1 - 4 μg/mg
Formulation PBS, pH 7.2, containing 40% glycerol with 0.02% sodium azide
Gene Alias sphingosine-1-phosphate phosphatase 1
Gene Symbols SGPP1
Host Species Rabbit
Immunogen This antibody was developed against a peptide corresponding to amino acids: YPFVDLIDNFNQTHKYAPFI
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 81537
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.