missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SGOL2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NB100-60454
This item is not returnable.
View return policy
Description
SGOL2 Polyclonal specifically detects SGOL2 in Human samples. It is validated for Immunoprecipitation, Western Blot (Negative).
Specifications
| SGOL2 | |
| Polyclonal | |
| Unconjugated | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
| FLJ25211, MGC129665, Sgo2, Shugoshin-2, shugoshin-like 2 (S. pombe), Tripin, TRIPINshugoshin-like 2 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 151246 | |
| Human | |
| IgG |
| Immunoprecipitation | |
| 1.0 mg/mL | |
| Immunoprecipitation 2-5 ug/mg lysate | |
| Q562F6 | |
| SGOL2 | |
| The immunogen recognized by this antibody maps to a region between residue 175 and 225 of human shugoshin-like 2 using the numbering given in entry NP_689737.3 (GeneID 151246). | |
| 100 μL | |
| Primary | |
| Human. Based on 100% sequence identity, this antibody is predicted to react with Rhesus Monkey, Gorilla, Chimpanzee, and Northern white-cheeked gibbon. | |
| Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction