missing translation for 'onlineSavingsMsg'
Learn More

SGLT3/SLC5A4 Antibody - BSA Free, Novus Biologicals™

Product Code. 18419424 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mL
Unit Size:
0.05mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18419424 0.05 mL 0.05mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18419424 Supplier Novus Biologicals Supplier No. NBP150164

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SGLT3/SLC5A4 Polyclonal antibody specifically detects SGLT3/SLC5A4 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen SGLT3/SLC5A4
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.88 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin 2.5-5 μg/mL
Formulation PBS
Gene Accession No. Q9NY91
Gene Alias DJ90G24.4, low affinity sodium-glucose cotransporter, Na(+)/glucose cotransporter 3, SAAT1low affinity sodium glucose cotransporter, SGLT2, SGLT3, sodium transporter, Sodium/glucose cotransporter 3, solute carrier family 5 (low affinity glucose cotransporter), member 4, solute carrier family 5 (neutral amino acid transporters, system A), member 4, Solute carrier family 5 member 4
Host Species Rabbit
Immunogen Synthetic 16 amino acid peptide from C-terminus of human SGLT3/SLC5A4.
Purification Method Immunogen affinity purified
Quantity 0.05 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6527
Target Species Human
Content And Storage Keep as concentrated solution. Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.