missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SGLT2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA559644
This item is not returnable.
View return policy
Description
Immunogen sequence: FHEVGGYSGL FDKYLGAATS LTVSEDPAVG NISSFCYRPR PDSYHLL Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 91%.
Sodium-dependent glucose transporter. Has a Na+ to glucose coupling ratio of 1:1. Efficient substrate transport in mammalian kidney is provided by the concerted action of a low affinity high capacity and a high affinity low capacity Na+/glucose cotransporter arranged in series along kidney proximal tubules.
Specifications
| SGLT2 | |
| Polyclonal | |
| Unconjugated | |
| SLC5A2 | |
| glucose transporter; bSGLT2; low affinity Na-dependent glucose transporter (SGLT2); low affinity sodium-dependent glucose cotransporter; low affinity sodium-glucose cotransporter; Na(+)/glucose cotransporter 2; Na(+)/nucleoside cotransporter; Na+/glucose cotransporter 2; Na+/nucleoside cotransporter; Sglt2; SGLT-2; SLC5A2; SNST1; sodium/glucose cotransporter 2; Sodium/nucleoside cotransporter; sodium-glucose cotransporter-like 2; solute carrier family 5 (sodium/glucose cotransporter), member 2; solute carrier family 5 (sodium/glucose transporter), member 2; solute carrier family 5 member 2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 6524 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.10 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P31639 | |
| SLC5A2 | |
| Recombinant protein corresponding to Human SGLT2. Recombinant protein control fragment (Product #RP-98643). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction