missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SGLT1 Recombinant Rabbit Monoclonal Antibody (9P7J4)

Rabbit Recombinant Monoclonal Antibody
£147.00 - £363.00
Specifications
| Antigen | SGLT1 |
|---|---|
| Clone | 9P7J4 |
| Concentration | 1.57 mg/mL |
| Content And Storage | -20°C, Avoid Freeze/Thaw Cycles |
| Applications | ELISA, Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30284028
|
Invitrogen™
MA557899 |
20 μL |
£147.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30283036
![]() |
Invitrogen™
MA557900 |
100 μL |
£363.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Immunogen sequence: MPMFIMVMPG MISRILYTEK IACVVPSECE KYCGTKVGCT NIAYPTLVVE LMPNGLRGLM LSVMLASLMS SLTSIFNSAS TLFTMDIYAK VRKRASEKEL.
Glucose transporters are integral membrane proteins that mediate the transport of glucose and structurally-related substances across cellular membranes. Two families of glucose transporter have been identified: the facilitated-diffusion glucose transporter family. The SLC5A1 gene encodes a protein that is involved in the active transport of glucose and galactose into eukaryotic and some prokaryotic cells.Specifications
| SGLT1 | |
| 1.57 mg/mL | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| P13866 | |
| 6523 | |
| Synthetic peptide. | |
| Antibody |
| 9P7J4 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human | |
| Slc5a1 | |
| D22S675; High affinity sodium-glucose cotransporter; HSGLT1; MGC93553; Na; Na(+)/glucose cotransporter 1; Na+/glucose cotransporter 1; NAGT; Sglt1; SLC5A1; sodium glucose cotransporter 1; sodium/glucose cotransporter 1; sodium-glucose cotransporter 1; sodium-glucose cotransporter-like 1; solute carrier family 5 (sodium/glucose cotransporter), member 1; solute carrier family 5 member 1; solute carrier family 5, member 1; solute carrier family 5, member alpha 1; Solute carrier family 5, member alpha 1 (Na+/glucose cotransporter) | |
| Slc5a1 | |
| Primary | |
| Affinity chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title