missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SGLT1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA588282
This item is not returnable.
View return policy
Description
Positive Samples: 293T, Hela, Mouse uterus; Cellular Location: Membrane, Multi-pass membrane protein Immunogen sequence: DIYAKVRKRA SEKEL.
Glucose transporters are integral membrane proteins that mediate the transport of glucose and structurally-related substances across cellular membranes. Two families of glucose transporter have been identified: the facilitated-diffusion glucose transporter family. The SLC5A1 gene encodes a protein that is involved in the active transport of glucose and galactose into eukaryotic and some prokaryotic cells.
Specifications
| SGLT1 | |
| Polyclonal | |
| Unconjugated | |
| Slc5a1 | |
| D22S675; High affinity sodium-glucose cotransporter; HSGLT1; MGC93553; Na; Na(+)/glucose cotransporter 1; Na+/glucose cotransporter 1; NAGT; Sglt1; SLC5A1; sodium glucose cotransporter 1; sodium/glucose cotransporter 1; sodium-glucose cotransporter 1; sodium-glucose cotransporter-like 1; solute carrier family 5 (sodium/glucose cotransporter), member 1; solute carrier family 5 member 1; solute carrier family 5, member 1; solute carrier family 5, member alpha 1; Solute carrier family 5, member alpha 1 (Na+/glucose cotransporter) | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 6523 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 1.01 mg/mL | |
| PBS with 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| P13866 | |
| Slc5a1 | |
| Synthetic peptide. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction