missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SGK494 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA567857
Additional Details : Weight : 0.01000kg
Description
Predicted to react with Mouse samples.
Component of the pupal cuticle.Specifications
SGK494 | |
Polyclonal | |
Unconjugated | |
SGK494 | |
sugen kinase 494 | |
Rabbit | |
Affinity chromatography | |
RUO | |
124923 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q96LW2 | |
SGK494 | |
Synthesized peptide derived from human SGK494. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |