Learn More
Invitrogen™ SGK2 Recombinant Human Protein
SGK2 is a serine/threonine protein kinase which is similar to serum- and glucocorticoid-induced protein kinase (SGK).
Brand: Invitrogen™ PR7313A
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Unlike SGK, SGK2 is not induced by serum or glucocorticoids. It is induced in response to signals that activate phosphatidylinositol 3-kinase, which is also true for SGK.
Specifications
NP_057360, S416D | |
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol | |
100μg | |
RUO - research use only | |
SGK2 | |
Human | |
GST Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
69.5kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Catalytic | |
SGK Kinase Family | |
Recombinant | |
Baculovirus |