missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SGK1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NBP2-19010
This item is not returnable.
View return policy
Description
SGK1 Polyclonal specifically detects SGK1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| SGK1 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.05% BSA with 0.05% Sodium Azide | |
| SGK1 | |
| A portion of amino acids 360-410 from human SGK1 was used as the immunogen for this antibody. NP_005618.2. | |
| 0.1 mg | |
| Apoptosis, DNA Repair | |
| 6446 | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 0.5 mg/ml | |
| Western Blot 1-3 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 5 ug/ml | |
| EC 2.7.11, EC 2.7.11.1, serine/threonine protein kinase SGK, serine/threonine-protein kinase Sgk1, serum/glucocorticoid regulated kinase, serum/glucocorticoid regulated kinase 1, Serum/glucocorticoid-regulated kinase 1, SGK | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| The amino acid sequence used as immunogen for this antibody is 100% homologous in human, new world monkey, macaque, rhesus monkey, opossum, gibbon, orangutan, and chimpanzee, 95% in panda, Carolina anole, cow, dog, Chinese hamster, zebrafish, horse, mole rat, elephant, platypus, and pig, 90% in mouse, rat, zebra finch, frog, cichlid fish, rabbit, guinea pig, chicken, and wild turkey, 85% homologouus in salmon. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction