missing translation for 'onlineSavingsMsg'
Learn More

Eurofins DiscoverX SGK1 (δ1-59, S422D) Protein, Active

Product Code. 17300144
Encompass_Preferred
Click to view available options
Quantity:
250 μg
6 μg
Unit Size:
250µg
6µg
This item is not returnable. View return policy

Product Code. 17300144

Brand: Eurofins DiscoverX 14331

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Active, N-truncated (missing N-terminal 60 amino acids), His-tagged, human SGK1 fusion protein, containing a S422D mutation, for use in Kinase Assays.

The serum/glucocorticoid regulated kinases (SGK1,2,3) are serine/threonine kinases that belong to the AGC (protein kinase A; protein kinase G and protein kinase C) subfamily that includes cAMP and cGMP dependent kinases, PKB and PKC. SGK1 (SGK) is predominately expressed in kidney, liver and pancreas. The catalytic domain of serum/glucocorticoid regulated protein kinase is most similar to PKB and activated via phosphorlation mechanism when cells are stimulated with insulin, IGF1 or serum, or exposed to oxidative stress.

GenBank AF153609. The protein is S422D with respect to this accession number.

Specifications

Formulation mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 150 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution.
Molecular Weight (g/mol) 45.7 kDa
Quantity 6 μg
Species Human
Recombinant Recombinant
Protein Tag N-terminal 6His
Expression System Recombinant enzyme expressed in Sf21 insect cells
Protein Form Truncated
Purity or Quality Grade ≥72%
Protein SGK1 (S422D), active
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.