missing translation for 'onlineSavingsMsg'
Learn More

SGI-7079, MedChemExpress

Product Code. p-200100007
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16491627 50 mg 50mg
16468167 5 mg 5mg
16456046 100 mg 100mg
16425458 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16491627 Supplier Medchem Express Supplier No. HY12964

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress SGI-7079 is a potent and ATP-competitive Axl inhibitor, significantly inhibits the proliferation of SUM149 or KPL-4 cells with an IC50 of 0.43 or 0.16 μM, respectively.

SGI-7079 is a potent and ATP-competitive Axl inhibitor, significantly inhibits the proliferation of SUM149 or KPL-4 cells with an IC50 of 0.43 or 0.16 μM, respectively.

Specifications

Chemical Name or Material SGI-7079
CAS 1239875-86-5
Color Yellow
Purity Grade Notes Research
Molecular Formula C26H26FN7
Quantity 50 mg
Solubility Information DMSO : ≥ 34 mg/mL (74.64 mM)
SMILES N#CCC1=CC=CC(C2=C3C(NC=C3C)=NC(NC4=CC=C(N5CCN(C)CC5)C(F)=C4)=N2)=C1
Molecular Weight (g/mol) 455.53
Formula Weight 455.53
Percent Purity 98.52%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.