missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SGI-7079, MedChemExpress
Description
SGI-7079 is a potent and ATP-competitive Axl inhibitor, significantly inhibits the proliferation of SUM149 or KPL-4 cells with an IC50 of 0.43 or 0.16 μM, respectively.
Specifications
Specifications
| Chemical Name or Material | SGI-7079 |
| CAS | 1239875-86-5 |
| Color | Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C26H26FN7 |
| Quantity | 50 mg |
| Solubility Information | DMSO : ≥ 34 mg/mL (74.64 mM) |
| SMILES | N#CCC1=CC=CC(C2=C3C(NC=C3C)=NC(NC4=CC=C(N5CCN(C)CC5)C(F)=C4)=N2)=C1 |
| Molecular Weight (g/mol) | 455.53 |
| Formula Weight | 455.53 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?