missing translation for 'onlineSavingsMsg'
Learn More

SGI-1776, MedChemExpress

Product Code. 16485618
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16485618 10 mg 10mg
16471787 50 mg 50mg
16448317 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16485618 Supplier Medchem Express Supplier No. HY13287
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress SGI-1776 is an inhibitor of Pim kinases, with IC50s of 7 nM, 363 nM, and 69 nM for Pim-1, -2 and -3, respectively.

SGI-1776 is an inhibitor of Pim kinases, with IC50s of 7 nM, 363 nM, and 69 nM for Pim-1, -2 and -3, respectively.

Specifications

Chemical Name or Material SGI-1776
CAS 1025065-69-3
Color White
Purity Grade Notes Research
Molecular Formula C20H22F3N5O
Quantity 10 mg
Solubility Information DMSO : 125 mg/mL (308.32 mM; Need ultrasonic)
SMILES CN1CCC(CC1)CNC2=NN3C(C=C2)=NC=C3C4=CC=CC(OC(F)(F)F)=C4
Molecular Weight (g/mol) 405.42
Formula Weight 405.42
Percent Purity 98.67%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.