missing translation for 'onlineSavingsMsg'
Learn More

SGI-1027, MedChemExpress

Product Code. 16445938
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16445938 10 mg 10mg
16442087 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16445938 Supplier Medchem Express Supplier No. HY13962
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress SGI-1027 is a DNA methyltransferase (DNMT) inhibitor, with IC50s of 7.5 μM, 8 μM, and 12.5 μM for DNMT3B, DNMT3A, and DNMT1 with poly(dI-dC) as substrate.

SGI-1027 is a DNA methyltransferase (DNMT) inhibitor, with IC50s of 7.5 μM, 8 μM, and 12.5 μM for DNMT3B, DNMT3A, and DNMT1 with poly(dI-dC) as substrate.

Specifications

Chemical Name or Material SGI-1027
CAS 1020149-73-8
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C27H23N7O
Quantity 10 mg
Solubility Information DMSO : 27 mg/mL (58.50 mM; Need ultrasonic and warming)
SMILES O=C(NC1=CC=C(NC2=NC(N)=NC(C)=C2)C=C1)C3=CC=C(NC4=CC=NC5=CC=CC=C45)C=C3
Molecular Weight (g/mol) 461.52
Formula Weight 461.52
Percent Purity 99.35%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.