missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SGEF Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94359-0.1ml
This item is not returnable.
View return policy
Description
SGEF Polyclonal antibody specifically detects SGEF in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| SGEF | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:100 | |
| Rho guanine nucleotide exchange factor (GEF) 26 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 592-871 of human ARHGEF26 (NP_056410.3). ICQKTPKDSPKYEVCKRALKEVSKLVRLCNEGARKMERTEMMYTINSQLEFKIKPFPLVSSSRWLVKRGELTAYVEDTVLFSRRTSKQQVYFFLFNDVLIITKKKSEESYNVNDYSLRDQLLVESCDNEELNSSPGKNSSTMLYSRQSSASHLFTLTVLSNHANEKVEMLLGAETQSERARWITALGHSSGKPPADRTSLTQVEIVRSFTAKQPDELSLQVADVVLIYQRVSDGWYEGERLRDGERGWFPMECAKEITCQATIDKNVERMGRLLGLETNV | |
| 0.1 mL | |
| GPCR, Signal Transduction | |
| 26084 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion