Learn More
Trajan™ Thermo Scientific Instrument Ferrules
Product Code. 10493392
Change view
Click to view available options
Diameter (Metric) Inside:
0.10 to 0.25 mm
0.32 mm
0.45 to 0.53 mm
Unit Size:
1 set
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Inside unitSize
10493392 0.32 mm Each
10072452 0.10 to 0.25 mm Each
12927101 0.45 to 0.53 mm 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10493392 Supplier Trajan™ Supplier No. 073451
Only null left
Add to basket
Add to basket
SilTite FingerTite delivers an easy, leak-free installation for capillary columns without the use of any tools
  • Next generation ferrule system for gas chromatography
  • Eliminates leaks
  • No contamination
  • No need to retighten after temperature cycling
  • Cleaner spectra for better library matches
Specifications
Type GC Interface Connection
Material SilTite Metal
Quantity 10 ferrules, 2 nuts
Diameter (Metric) Inside 0.32 mm