Learn More
Trajan™ Thermo Scientific AS3000, AS2000 and AS200/800GC Autosampler Syringe
Fixed needle autosampler syringes. For reduced non specific sample interaction and increased analyte recovery
Brand: Trajan™ 0029801
Code : A0
Additional Details : Weight : 0.20000kg
Description
- High quality solutions for liquid handling
- Accurate and reproducible injections
- Specifically designed for Thermo Scientific instruments
Specifications
26 G | |
AS3000, AS2000 and AS200/800GC Autosampler Syringe | |
10 μL |
0.47 mm | |
1/Pk. |