Learn More
SGE™ SilTite™ FingerTite Ferrule System Starter Kits
Product Code. 15268943
Change view
Click to view available options
For Use With:
Agilent Capillary/FID
Agilent INJ/FID
PerkinElmer INJ/FID
PerkinElmer INJ/MS
SGE SilTite
Shimadzu 2010 INJ/FID
Shimadzu 2010 INJ/MS
Shimadzu Agilent INJ/FID
Shimadzu Agilent INJ/MS
Thermo INJ/DSQ/DSQII
Thermo INJ/ISQ
Varian/Bruker INJ/FID
Varian/Bruker INJ/MS
Unit Size:
Each
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With unitSize
15268943 Shimadzu Agilent INJ/FID Each
15218973 Shimadzu 2010 INJ/MS Each
10386458 Shimadzu Agilent INJ/MS Each
15288963 Shimadzu 2010 INJ/FID Each
15288943 SGE SilTite Each
15238953 SGE SilTite Each
15248953 Agilent INJ/FID Each
16632642 Agilent Capillary/FID Each
15228963 Thermo INJ/ISQ Each
15238963 Thermo INJ/DSQ/DSQII Each
15248963 Varian/Bruker INJ/FID Each
15278963 Varian/Bruker INJ/MS Each
15228973 PerkinElmer INJ/FID Each
15258973 PerkinElmer INJ/MS Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Product Code. 15268943 Supplier SGE™ Supplier No. 073610
Only null left
Add to basket
Add to basket
For gas chromatography systems
SilTite FingerTite is the next generation ferrule system for gas chromatography systems delivering an easy, leak free installation for capillary columns without the use of any tools. Each starter kit includes all the parts necessary to convert one GC system (one injector and one detector) to the SilTite FingerTite system. The system requires no permanent instrument modifications—switching back to conventional connections can be done quickly and easily.
  • Easy to use
  • Reliable connecton
  • Leak free every time
  • Removes contamination caused by Vespel™ or graphite ferrules
  • No need to re-tighten ferrules after temperature cycling
  • Reduce air background
  • NO TOOLS (FingerTite)
Specifications
Type FingerTite Ferrule Starter Kit
For Use With Shimadzu Agilent INJ/FID