Learn More
SGE™ SilTite™ Ferrule Starter Kits Replacement SilTite Nuts and Base Seals
Specially designed ferrule and nut system developed specifically for fused silica GC columns and GC/MS systems
Brand: SGE™ 073401
Code : 13
Additional Details : Weight : 0.10000kg
Description
For use with SGE SilTite Ferrule Starter Kits
SilTite Nuts
- Non-Swagelok™ nuts with two notches on outer surface
- Inner taper of the nut corresponds to angle of ferrule, ensuring an even closure of the ferrule around the whole column to eliminate leaks
- Ferrules are easily removed from nut while still attached to capillary column
- Reusable
Specifications
SilTite Metal Ferrule | |
Split/Splitless Injector | |
Inlet Gold Base Seal |
Compatible with Agilent Split /Splitless Injector | |
Metal | |
10/Pk. |