Learn More
SGE™ SilTite™ Ferrule Starter Kits Replacement SilTite Nuts and Base Seals
Click to view available options
Type:
Inlet Gold Base Seal
Nut
Quantity:
10/Pk.
2/Pk.
5/Pk.
Unit Size:
Each
Description
For use with SGE SilTite Ferrule Starter Kits
SilTite Nuts
- Non-Swagelok™ nuts with two notches on outer surface
- Inner taper of the nut corresponds to angle of ferrule, ensuring an even closure of the ferrule around the whole column to eliminate leaks
- Ferrules are easily removed from nut while still attached to capillary column
- Reusable
Specifications
Specifications
| Type | Nut |
| For Use With (Application) | Split/Splitless Injector |
| For Use With (Equipment) | Split/Splitless Injectors |
| Material | Metal |
| Quantity | 5/Pk. |