Learn More
SGE™ SilFlow™ GC Replacement Parts
Tightened using finger force to achieve a reliable seal, even for sensitive MS systems
Brand:  SGE™ 123704
Code : NEW
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15289163
  • £267.00 / Pack of 10
Estimated Shipment: 31-05-2024
to see stock.
Add to basket
Description
Description
  • Conventional graphite or Vespel™ ferrules cannot be used with the SilFlow MCD as the dimensional stability is not adequate and there is a risk of contaminating the channels
  • SilTite™ FingerTite metal ferrules result in a reliable zero dead volume connection
Specifications
Specifications
SGE Silflow Microchannel Device
Nut
Nut