Learn More
SGE™ SilFlow™ GC 3 Port Splitter Kits
Product Code. 15289173
Change view
Click to view available options
Diameter (Metric) Inner:
0.25/0.32 mm
0.53 mm
Port B and C 0.25/0.32 mm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Inner unitSize
15289173 Port B and C 0.25/0.32 mm Each
12700948 0.53 mm Each
13589000 0.25/0.32 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15289173 Supplier SGE™ Supplier No. 123720
Only null left
Add to basket
Add to basket
Highly efficient and reliable micro fluidic platform to improve GC connectivity.
  • Kit includes a SilFlow microchannel device (MCD), SilTite FingerTite tool, mounting bracket, plus appropriate ferrules, nuts and blanking ferrules for set up
  • 1.1mm OD tubing not included
Specifications
Type Port Splitter
Diameter (Metric) Outer 1.1 mm
Diameter (Metric) Inner Port B and C 0.25/0.32 mm
Includes SilFlow microchannel device (MCD), SilTite FingerTite tool, Mounting bracket, Appropriately sized ferrules, nuts and blanking ferrules for set up
For Use With SGE Silflow Microchannel Device