Learn More
Trajan™ Replacement Plungers for Diamond Manual Syringes
Product Code. 13167956
Click to view available options
For Use With (Equipment):
Diamond Manual Syringe (10μL)
Diamond Manual Syringe (100μL)
Diamond Manual Syringe (25μL)
Diamond Manual Syringe (250μL)
Diamond Manual Syringe (50μL)
Diamond Manual Syringe (500μL)
Unit Size:
Each
Product Code. 13167956
Brand: Trajan™ 0355102
SGE syringe replacement plungers for gas tight syringes
  • Replacement plungers for gas tight syringes
  • Designed to match the specific syringe barrel to maintain a gas tight seal
Specifications
Material PTFE (Plunger)
Product Type Replacement Plunger
For Use With (Equipment) Diamond Manual Syringe (25μL)