Learn More
Trajan™ Replacement Needles For 5ul Removable Needle Syringes
Product Code. 15208593
Change view
Click to view available options
Needle Gauge:
23 G
Unit Size:
Pack of 2
Product Code. Needle Gauge unitSize
15208593 23 G Pack of 2
1 options
Product Code. 15208593 Supplier Trajan™ Supplier No. 036011
Only null left
Add to basket
Add to basket
Quality needles to fit SGE syringes
  • Needle length is exposed length of needle
  • Choose different replacement needle by selecting syringe volume, needle length, OD and tip style
Specifications
Needle Gauge 23 G
Diameter (Metric) Inner Needle 0.11 mm
Diameter (Metric) Outer Needle 0.63 mm
Needle Point Style Cone