missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trajan™ PerkinELmer Split Straight-Through Liner
The unique deactivation process of SGE inlet liners provides excellent analysis for a wide range of compounds
Brand: Trajan™
Code : 13
Additional Details : Weight : 0.50000kg
Description
- Each batch of liners tested for activity
- Manufactured from borosilicate glass
- Color coded by geometry for easy identification
- For use with PerkinElmer instruments
Specifications
GC Inlet liner, Straight Through | |
92 mm | |
6.2 mm |
5/Pk. | |
4 mm |