missing translation for 'onlineSavingsMsg'
Learn More

Trajan™ PerkinElmer AutoSystem and Clarus 500 Liners

The unique deactivation process of SGE inlet liners provides excellent analysis for a wide range of compounds

Brand:  Trajan™ 09224401

Code : 13

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 12944081

  • £71.00 / Each
Estimated Shipment: 24-06-2024
to see stock.
Add to basket

Description

Description

  • Each batch of liners tested for activity
  • Manufactured from borosilicate glass
  • Color coded by geometry for easy identification
  • For use with PerkinElmer instruments
Specifications

Specifications

AutoSystem and Clarus 500 liner, Large Volume Injection
86.2 mm
4 mm
Sintered Glass
1 Ea.
2 mm
AutoSystem XL (PSS Injector)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers