missing translation for 'onlineSavingsMsg'
Learn More

SGE™ GC Septa: CS

Product Code. 10390321
Change view
Click to view available options
Diameter (English) Inner:
0.18 in.
0.37 in.
0.39 in.
0.43 in.
0.45 in.
0.5 in.
0.59 in.
Unit Size:
Pack of 48
Pack of 50
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (English) Inner unitSize
10390321 0.37 in. Pack of 50
10095260 0.39 in. Pack of 50
10106830 0.45 in. Pack of 50
10472541 0.59 in. Pack of 48
10742572 0.18 in. Pack of 50
10055430 0.43 in. Pack of 50
17128015 0.5 in. Pack of 48
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 10390321 Supplier SGE™ Supplier No. 0418240

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Designed specifically for use in Agilent Technologies™ (HP) GC instruments

CS Septa

  • PTFE-coated silicone septa for low temperature applications
  • Good durability, good resealing
  • Excellent solvent resistance; good tear resistance
  • Maximum temperature: 200°C (392°F)

Specifications

Diameter (English) Inner 0.37 in.
Material PTFE-coated Silicone
Quantity 50/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.