missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SGE™ GC Inlet Liner Accessories
Description
- Viton™ O-Ring—Can be used up to temperatures of 300°C (572°F)
- Graphite Sealing Rings—Can be used up to temperatures of 450°C (842°F)
Specifications
Specifications
| Type | SGE GC Inlet Liner Accessory |
| For Use With (Application) | Graphite Sealing Ring |
| For Use With (Equipment) | Varian 1078, 1079 Injectors |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?