Learn More
Trajan™ SilTite™ GC Capillary Column Connectors: μ Unions
Description
- Low thermal mass
- Zero Dead Volume
- Inert
- No-need to retighten after installation
- Connect Pre-column or GC x GC
Specifications
Specifications
| Type | Connector |
| Connections | 0.32 mm and 0.32 mm |
| Includes | 2 unions, 5 double taper ferrules and tools |
| For Use With (Equipment) | GC Capillary Column |