Learn More
Trajan™ SilTite™ GC Capillary Column Connectors: μ Unions
Product Code. 15248933
Change view
Click to view available options
Connections:
0.1-0.25 mm and 0.1-0.25 mm
0.1-0.25 mm and 0.32 mm
0.1-0.25 mm and 0.53 mm
0.32 mm and 0.32 mm
0.32 mm and 0.53 mm
0.53 mm and 0.53 mm
Unit Size:
Each
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Connections unitSize
15248933 0.1-0.25 mm and 0.32 mm Each
15258933 0.32 mm and 0.32 mm Each
11353662 0.1-0.25 mm and 0.53 mm Each
12736368 0.1-0.25 mm and 0.1-0.25 mm Each
16692632 0.1-0.25 mm and 0.1-0.25 mm Each
15268933 0.32 mm and 0.53 mm Each
15278933 0.53 mm and 0.53 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 15248933 Supplier Trajan™ Supplier No. 073561
Only null left
Add to basket
Add to basket
SilTite μ-Union delivers an easy, leak-free installation for capillary columns without the use of any tools
  • Low thermal mass
  • Zero Dead Volume
  • Inert
  • No-need to retighten after installation
  • Connect Pre-column or GC x GC
Specifications
Type Connector
Connections 0.1-0.25 mm and 0.32 mm
Includes 2 unions, 5 double taper ferrules and tools
For Use With (Equipment) GC Capillary Column