Learn More
SGE™ BP20 (Wax) GC Capillary Columns
Combines innovative phase technology and unique fused silica production capability. SGE™ BP20 (Wax) GC Capillary Columns provides end-to-end separation solutions for all applications.
£303.00 - £1025.00
Specifications
Stationary Phase SGE™ BP20 (Wax)
USP Type G14, G15, G16, G20, G39
Functionality/Interaction Type Hydrogen Bonding or Dipole interactions
Polarity Polar
Coating Fused Silica
View More Specs
Products 21
Product Code Brand Diameter (Metric) Inner Film Thickness Length (Metric) Price Quantity & Availability  
Product Code Brand Diameter (Metric) Inner Film Thickness Length (Metric) Price Quantity & Availability  
15238673
View Documents Promotion Details
SGE™
054424
0.22 mm 0.25 μm 30 m
£530.00
Each
View alternative products
Save Up to 
»
Estimated Shipment: 23-05-2024
to see stock.
Add to basket
10742372
View Documents Promotion Details
SGE™
054431
0.32 mm 0.25 μm 50 m
£980.00
Each
View alternative products
Save Up to 
»
Estimated Shipment: 23-05-2024
to see stock.
Add to basket
12985581
View Documents Promotion Details
SGE™
054437
0.32 mm 0.5 μm 50 m
£980.00
Each
View alternative products
Save Up to 
»
Estimated Shipment: 23-05-2024
to see stock.
Add to basket
15208683
View Documents Promotion Details
SGE™
054443
0.32 mm 1 μm 50 m
£980.00
Each
View alternative products
Save Up to 
»
Estimated Shipment: 23-05-2024
to see stock.
Add to basket
15225263
View Documents Promotion Details
SGE™
054456
0.53 mm 2 μm 25 m
£620.00
Each
View alternative products
Save Up to 
»
Estimated Shipment: 23-05-2024
to see stock.
Add to basket
16682612
View Documents Promotion Details
SGE™
054456
0.53 mm 2 μm 25 m
£620.00
Each
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Add to basket
12915591
View Documents Promotion Details
SGE™
054405
0.1 mm 0.1 μm 10 m
£303.00
Each
View alternative products
Save Up to 
»
Estimated Shipment: 23-05-2024
to see stock.
Add to basket
10360751
View Documents Promotion Details
SGE™
054421
0.22 mm 0.25 μm 25 m
£530.00
Each
View alternative products
Save Up to 
»
Estimated Shipment: 04-06-2024
to see stock.
Add to basket
15228673
View Documents Promotion Details
SGE™
054415
0.25 mm 0.5 μm 30 m
£550.00
Each
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Add to basket
15248673
View Documents Promotion Details
SGE™
054425
0.22 mm 0.25 μm 60 m
£910.00
Each
View alternative products
Save Up to 
»
Estimated Shipment: 23-05-2024
to see stock.
Add to basket
10665441
View Documents Promotion Details
SGE™
054422
0.22 mm 0.25 μm 50 m
£910.00
Each
View alternative products
Save Up to 
»
Estimated Shipment: 23-05-2024
to see stock.
Add to basket
12674955
View Documents Promotion Details
SGE™
054436
0.32 mm 0.5 μm 25 m
£565.00
Each
View alternative products
Save Up to 
»
Estimated Shipment: 03-05-2024
to see stock.
Add to basket
12975581
View Documents Promotion Details
SGE™
054434
0.32 mm 0.25 μm 60 m
£955.00
Each
View alternative products
Save Up to 
»
Estimated Shipment: 23-05-2024
to see stock.
Add to basket
15288673
View Documents Promotion Details
SGE™
054440
0.53 mm 0.5 μm 30 m
£620.00
Each
View alternative products
Save Up to 
»
Estimated Shipment: 23-05-2024
to see stock.
Add to basket
15298673
View Documents Promotion Details
SGE™
054442
0.32 mm 1 μm 25 m
£565.00
Each
View alternative products
Save Up to 
»
Estimated Shipment: 23-05-2024
to see stock.
Add to basket
15238683
View Documents Promotion Details
SGE™
054445
0.32 mm 1 μm 60 m
£1025.00
Each
View alternative products
Save Up to 
»
Estimated Shipment: 29-05-2024
to see stock.
Add to basket
12905591
View Documents Promotion Details
SGE™
054450
0.53 mm 1 μm 15 m
£411.00
Each
View alternative products
Save Up to 
»
Estimated Shipment: 03-05-2024
to see stock.
Add to basket
15218683
View Documents Promotion Details
SGE™
054444
0.53 mm 1 μm 30 m
£565.00
Each
View alternative products
Save Up to 
»
Estimated Shipment: 23-05-2024
to see stock.
Add to basket
10157390
View Documents Promotion Details
SGE™
054448
0.53 mm 1 μm 25 m
£620.00
Each
View alternative products
Save Up to 
»
Estimated Shipment: 23-05-2024
to see stock.
Add to basket
15258683
View Documents Promotion Details
SGE™
054457
0.32 mm 0.5 μm 60 m
£980.00
Each
View alternative products
Save Up to 
»
Estimated Shipment: 23-05-2024
to see stock.
Add to basket
12766557
View Documents Promotion Details
SGE™
054458
0.25 mm 0.5 μm 60 m
£925.00
Each
View alternative products
Save Up to 
»
Estimated Shipment: 22-05-2024
to see stock.
Add to basket
Description
Description
Features
  • Industry standard Wax column
  • Bonded and cross-linked
  • Can be solvent rinsed
Applications
  • Used for separation of Alcohol, Free acids, Fatty acids methyl esters, Aromatics, Solvents and Essential oils
Specifications
Specifications
SGE™ BP20 (Wax)
Hydrogen Bonding or Dipole interactions
Fused Silica
G14, G15, G16, G20, G39
Polar