Learn More
Trajan™ Hexnut Fittings
Product Code. 15249103
Change view
Click to view available options
Includes:
10 hexnuts and 10 ferrules
2 hexnuts and 2 ferrules
Unit Size:
Pack of 10
Pack of 2
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes unitSize
15249103 2 hexnuts and 2 ferrules Pack of 2
15259103 10 hexnuts and 10 ferrules Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15249103 Supplier Trajan™ Supplier No. 1021003
Estimated Shipment: 18-09-2026
Add to basket
Add to basket
HexNut fittings provide ideal connections in HPLC systems
  • Stainless steel 10-32 thread
  • Inert contact surface makes it biocompatible
  • Ideal for use with corrosive solvents
Specifications
Includes 2 hexnuts and 2 ferrules
Thread Size 10-32
Material Stainless Steel
Pressure 5000 psi