Learn More
Trajan™ Methyl Deactivated Fused Silica Capillary Tubing
Essential for maintaining a high performance chromatography system.Trajan Scientific and Medical™ Methyl Deactivated Fused Silica Capillary Tubing is chemically inert and thermally stable up to 380°C.
Brand: Trajan™ 0624450
Code : NEW
Additional Details : Weight : 0.01000kg
Description
- Suitable for organic and aqueous solvents
- Methyl deactivated for use with hydrocarbons (pentane, hexane, heptane, iso-octane, aromatics and mixed solvents - n-paraffins/chlorinated solvents
- Individually tested performance of each 30m length from which all shorter lengths are prepared
- Ideal for biotechnology applications
Specifications
Capillary Tubing | |
0.002 in. | |
0.075 mm | |
6.56 ft. | |
Fused Silica | |
380°C |
Ideal for Biotechnology Applications, and Organic and Aqueous Solvents | |
0.007 in. | |
0.19 mm | |
2 m | |
716°F |