Learn More
Trajan™ CTC PAL HPLC Fixed Needle Autosampler Syringe
Product Code. 12967526
Change view
Click to view available options
Volume (Metric):
25 μL
100 μL
500 μL
Model:
100F-CTC-GT-LC
100F-CTC-GT-LC(0.4)
25F-CTC-GT-LC
500F-CTC-GT-LC(0.4)
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Model unitSize
12967526 100 μL 100F-CTC-GT-LC(0.4) Each
12947556 100 μL 100F-CTC-GT-LC Each
12987526 500 μL 500F-CTC-GT-LC(0.4) Each
12957526 25 μL 25F-CTC-GT-LC Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 12967526 Supplier Trajan™ Supplier No. 005720
Only null left
Add to basket
Add to basket
Borosilicate glass barrel and stainless steel needles provide strength and inertness. Backing provides quick and accurate volume assessment.
  • High temperature syringes for headspace application with temperatures up to 150°C
  • PTFE plunger seal option for smooth, leak free operation - suitable for both liquid and gas samples, with replaceable plungers
  • Specifications of the syringes are exactly matched to CTC holders to ensure proper fit and reliable functioning
Specifications
Needle Gauge 22 G
Diameter (Metric) Outer 0.72 mm
Product Type Fixed Needle Syringe
Model 100F-CTC-GT-LC(0.4)
Volume (Metric) 100 μL
Length (Metric) Needle 51 mm
Needle Point Style LC