Learn More
Trajan™ 5uL Plunger-in-Needle Syringes
Product Code. 10441401
Change view
Click to view available options
Volume (Metric):
5 μL
Unit Size:
Each
Product Code. Volume (Metric) unitSize
10441401 5 μL Each
1 options
Product Code. 10441401 Supplier Trajan™ Supplier No. 000850
Only null left
Add to basket
Add to basket
Designed to provide high precision and accuracy for small volume injections.
  • High quality solutions for liquid handling
  • Accurate and reproducible injections
  • 8mm OD barrel with Repeating Adaptor
Specifications
Needle Gauge 23 G
Diameter (Metric) Outer Needle 0.63 mm
Volume (Metric) 5 μL
Needle Point Style Cone