Learn More
Invitrogen™ SGCG Recombinant Rabbit Monoclonal Antibody (JA11-57)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA532671
Description
Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
Component of the sarcoglycan complex, a subcomplex of the dystrophin-glycoprotein complex which forms a link between the F-actin cytoskeleton and the extracellular matrix.
Specifications
| SGCG | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| TBS with 0.05% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4 | |
| P82348, Q13326 | |
| Sgcg | |
| Recombinant protein within Human gamma Sarcoglycan aa 42-291. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| JA11-57 | |
| Unconjugated | |
| Sgcg | |
| 35 kDa dystrophin-associated glycoprotein; 35DAG; 35kD dystrophin-associated glycoprotein; 35kDa; 5430420E18Rik; A4; AI642964; DAGA4; DMDA; DMDA1; gamma sarcoglycan; gamma-sarcoglycan; gamma-SG; LGMD2C; MAM; sarcoglycan gamma; sarcoglycan, gamma; sarcoglycan, gamma (35kD dystrophin-associated glycoprotein); sarcoglycan, gamma (35kDa dystrophin-associated glycoprotein); sarcoglycan, gamma (dystrophin-associated glycoprotein); SCARMD2; SCG3; SGCG; sgcg gene for gamma-sarcoglycan; TYPE; zgc:100924 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 24053, 6445 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.