missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SGCG Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18102-1-AP-150UL
This item is not returnable.
View return policy
Description
Component of the sarcoglycan complex, a subcomplex of the dystrophin-glycoprotein complex which forms a link between the F-actin cytoskeleton and the extracellular matrix.Specifications
| SGCG | |
| Polyclonal | |
| Unconjugated | |
| SGCG | |
| 35DAG, A4, DAGA4, DMDA, DMDA1, Gamma sarcoglycan, Gamma SG, LGMD2C, MAM, SCARMD2, SCG3, SGCG, TYPE | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 24053, 305941, 6445 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.15 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P82348, Q13326 | |
| SGCG | |
| SGCG Fusion Protein Ag12402 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction