missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SGCE Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA567363
This item is not returnable.
View return policy
Description
Immunogen sequence: DIQLVHHSAIQ KSTKELRDMS KNREIAWPLS TLPVFHPVTG EIIPPLHTDN YDSTNMPLMQ TQQNLPHQTQ IPQQQTTGKW Y Highest antigen sequence identity to the following orthologs - mouse 94%, rat 96%.
This gene encodes the epsilon member of the sarcoglycan family. Sarcoglycans are transmembrane proteins that are components of the dystrophin-glycoprotein complex, which link the actin cytoskeleton to the extracellular matrix. Unlike other family members which are predominantly expressed in striated muscle, the epsilon sarcoglycan is more broadly expressed. Mutations in this gene are associated with myoclonus-dystonia syndrome. This gene is imprinted, with preferential expression from the paternal allele. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications
| SGCE | |
| Polyclonal | |
| Unconjugated | |
| SGCE | |
| dystonia 11, myoclonic; DYT11; EMBL:AAO37579.1, ECO:0000312; EMBL:AAO37579.1}; Epsilon sarcoglycan; Epsilon SG; Epsilon-sarcoglycan; epsilon-sarcoglycan {ECO:0000312; epsilon-SG; epsilon-SG {ECO:0000250; ESG; e-SG; RGD:1303201}; sarcoglycan; sarcoglycan epsilon; sarcoglycan, epsilon; SGCE; sgce {ECO:0000312; UniProtKB:O43556}; UNQ433/PRO840 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8910 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O43556 | |
| SGCE | |
| Recombinant Human SGCE. Recombinant protein control fragment (Product #RP-105591). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction