Learn More
Invitrogen™ SGCB Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5110363
Description
Immunogen sequence: PNGCDSMEFH ESGLLRFKQV SDMGVIHPLY KSTVGGRRNE NLVITGNNQP IVFQQGTTKL SVENNKTSIT SDIGMQFFDP RTQNILFSTD YETHEFHLPS GVKSLNVQKA STERITSNAT SDLNIKVDGR AIVRGNEGVF IMGKTIEFHM GGNMELKAEN SIILNGSVMV STTRLPSSSS GDQLGSGDWV RYKLCMCADG TLFKVQVTSQ NMGCQISDNP CGNTH.
The function of this protein remains unknown.
Specifications
| SGCB | |
| Polyclonal | |
| Unconjugated | |
| SGCB | |
| 43 kDa dystrophin-associated glycoprotein; 43DAG; 43kDa; A3b; AI747103; AI844814; beta sarcoglycan; beta-sarcoglycan; beta-sarcoglycan(43kD dystrophin-associated glycoprotein); beta-SG; fa03a05; LGMD2E; limb girdle muscular dystrophy 2E (non-linked families); sarcoglycan beta; sarcoglycan, beta (43kD dystrophin-associated glycoprotein); sarcoglycan, beta (43kDa dystrophin-associated glycoprotein); sarcoglycan, beta (dystrophin-associated glycoprotein); SGC; Sgcb; wu:fa03a05; zgc:110833 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 24051, 6443, 680229 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.33 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P82349, Q16585 | |
| SGCB | |
| Recombinant protein (or fragment) | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.