missing translation for 'onlineSavingsMsg'
Learn More

SGC707, MedChemExpress

Product Code. 16487858
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16487858

Brand: Medchem Express HY19715

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress SGC707 is a first-in-class PRMT3 chemical probe which is a potent, selective, and cell-active allosteric inhibitor of PRMT3 with IC50 of 31 nM.

SGC707 is a first-in-class PRMT3 chemical probe which is a potent, selective, and cell-active allosteric inhibitor of PRMT3 with IC50 of 31 nM.

Specifications

Chemical Name or Material SGC707
Physical Form Solid
CAS 1687736-54-4
Color Light Yellow
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C16H18N4O2
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : ≥ 100 mg/mL (335.19 mM)
SMILES O=C(NCC(N1CCCC1)=O)NC2=CC=C3C=NC=CC3=C2
Molecular Weight (g/mol) 298.34
Formula Weight 298.34
Percent Purity 99.39%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.