missing translation for 'onlineSavingsMsg'
Learn More

SGC3027, MedChemExpress

Product Code. 16401179
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16401179 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16401179 Supplier Medchem Express Supplier No. HY112445
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress SGC3027 is a histone methyltransferase inhibitor. SGC3027 is the first potent, selective and cell active chemical probe for PRMT7.

SGC3027 is a histone methyltransferase inhibitor. SGC3027 is the first potent, selective and cell active chemical probe for PRMT7.

Specifications

Chemical Name or Material SGC3027
CAS 2624313-13-7
Color Yellow
Purity Grade Notes Research
Molecular Formula C41H47ClN6O6S
Quantity 1 mg
Solubility Information DMSO : 250 mg/mL (317.51 mM; Need ultrasonic)
SMILES ClC1=CC=C(C2=CC(CN(C(CC(C)(C)C3=C(C)C(C(C)=C(C)C3=O)=O)=O)CCCCSC[C@@H]4[C@@H](O)[C@@H](O)[C@H](N5C(N=CN=C6N)=C6N=C5)O4)=CC=C2)C=C1
Molecular Weight (g/mol) 787.37
Formula Weight 787.37
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.