missing translation for 'onlineSavingsMsg'
Learn More

SGC2085, MedChemExpress

Product Code. 16414386
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16414386 100 mg 100mg
16470669 1 mg 1mg
16455817 5 mg 5mg
16439646 50 mg 50mg
16413088 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16414386 Supplier Medchem Express Supplier No. HY100565

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress SGC2085 is a potent and selective coactivator associated arginine methyltransferase 1 (CARM1) inhibitor with an IC50 of 50 nM.

SGC2085 is a potent and selective coactivator associated arginine methyltransferase 1 (CARM1) inhibitor with an IC50 of 50 nM.

Specifications

Chemical Name or Material SGC2085
CAS 1821908-48-8
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C19H24N2O2
Quantity 100 mg
Solubility Information DMSO : ≥ 32 mg/mL (102.43 mM)
SMILES CC1=CC(OC2=CC=C(CNC([C@@H](N)C)=O)C=C2C)=CC(C)=C1
Molecular Weight (g/mol) 312.41
Formula Weight 312.41
Percent Purity 99.45%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.