missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trajan Scientific and Medical™ Varian Split / Splitless FocusLiner
The unique deactivation process of SGE inlet liners provides excellent analysis for a wide range of compounds
Brand: Trajan Scientific and Medical™ 092037
Code : 13
Additional Details : Weight : 0.01000kg
Description
- Each batch of liners tested for activity
- Manufactured from borosilicate glass
- Color coded by geometry for easy identification
- For use with Thermo Scientific instruments
Specifications
GC Inlet Liner | |
54 mm | |
5 mm |
5/Pk. | |
3.4 mm |