missing translation for 'onlineSavingsMsg'
Learn More

SGC-iMLLT, MedChemExpress

Product Code. 16455536
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16455536

Brand: Medchem Express HY112804

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress SGC-iMLLT is a first-in-class chemical probe and a potent, selective inhibitor of MLLT1/3-histone interactions with an IC50 of 0.26 μM. SGC-iMLLT shows high binding activity towards MLLT1 YEATS domain (YD) and MLLT3 YD (AF9/YEATS3) with Kds of 0.129 and 0.077 μM, respectively.

SGC-iMLLT is a first-in-class chemical probe and a potent, selective inhibitor of MLLT1/3-histone interactions with an IC50 of 0.26 μM. SGC-iMLLT shows high binding activity towards MLLT1 YEATS domain (YD) and MLLT3 YD (AF9/YEATS3) with Kds of 0.129 and 0.077 μM, respectively.

Specifications

Chemical Name or Material SGC-iMLLT
Physical Form Powder
CAS 2255338-25-9
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C22H24N6O
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : 83.33 mg/mL (214.51 mM; Need ultrasonic)
SMILES CN1N=CC2=CC(C(NC3=CC=C(NC(CN4[C@@H](C)CCC4)=N5)C5=C3)=O)=CC=C21
Molecular Weight (g/mol) 388.47
Formula Weight 388.47
Percent Purity 95.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.