missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ SGC GAK 1
High affinity cyclin G associated kinase (GAK) inhibitor
£83.15 - £314.25
Specifications
Chemical Name or Material | 6-Bromo-N-(3,4,5-trimethoxyphenyl)-4-quinolinamine |
---|---|
CAS | 2226517-76-4 |
Molecular Formula | C18H17BrN2O3 |
Purity | 99% |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18708838
|
Tocris Bioscience™
6564/5 |
5 mg |
£83.15
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18715348
|
Tocris Bioscience™
6564/25 |
25 mg |
£314.25
25mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
High affinity cyclin G associated kinase (GAK) inhibitor (Kd = 1.9 nM). Exhibits >30-fold selectivity for GAK over a panel of other kinases. Inhibits GAK in live cell assays (IC50 = 110 nM).Specifications
6-Bromo-N-(3,4,5-trimethoxyphenyl)-4-quinolinamine | |
2226517-76-4 | |
C18H17BrN2O3 | |
99% |
Safety and Handling
Recommended Storage : Store at -20°C