missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ SGC-CBP30
Description
Potent CBP/p300 bromodomain (BRD) inhibitor (Kd values are 21 and 32 nM for CBP and p300 BRDs respectively). Exhibits 40-fold and 250-fold selectivity for CBP over the first BRD of BRD4 (BRD4(1)) and BRD4(2) respectively. Accelerates FRAP recovery in cells at a concentration of 1 μM.
Specifications
Specifications
| Quantity | 50 mg |
| Formula Weight | 509.04 |
| Purity | >98% |
| Molecular Weight (g/mol) | 509.05 |
| Chemical Name or Material | SGC-CBP30 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?