missing translation for 'onlineSavingsMsg'
Learn More

SGC-CBP30, MedChemExpress

Product Code. 16496848
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16496848 10 mg 10mg
16482917 50 mg 50mg
16429397 5 mg 5mg
16416956 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16496848 Supplier Medchem Express Supplier No. HY15826

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress SGC-CBP30 is a potent and highly selective CBP/p300 bromodomain (Kds of 21 nM and 32 nM for CBP and p300, respectively) inhibitor, displaying 40-fold selectivity over the first bromodomain of BRD4 [BRD4(1)] bound. SGC-CBP30 strongly reduces secretion of IL-17A in Th17 cells and has anti-inflammatory effects.

SGC-CBP30 is a potent and highly selective CBP/p300 bromodomain (Kds of 21 nM and 32 nM for CBP and p300, respectively) inhibitor, displaying 40-fold selectivity over the first bromodomain of BRD4 [BRD4(1)] bound. SGC-CBP30 strongly reduces secretion of IL-17A in Th17 cells and has anti-inflammatory effects.

Specifications

Chemical Name or Material SGC-CBP30
CAS 1613695-14-9
Color Yellow
Purity Grade Notes Research
Molecular Formula C28H33ClN4O3
Quantity 10 mg
Solubility Information DMSO : ≥ 31 mg/mL (60.90 mM)
SMILES C[C@H](N1CCOCC1)CN2C(CCC3=CC=C(OC)C(Cl)=C3)=NC4=CC(C5=C(C)ON=C5C)=CC=C42
Molecular Weight (g/mol) 509.04
Formula Weight 509.04
Percent Purity 99.06%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.