missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ SGC AAK1 1

Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
Description
SGC AAK1 1 is an adaptor-associated protein kinase 1 (AAK1) and BMP-2-inducible protein kinase (BMP2K) inhibitor (IC50 values are 233 nM and 1.48 μM for AAK1 and BMP2K, respectively). Inhibits AAK1-dependent phosphorylation of AP2M1. Activates Wnt signaling.
Specifications
Specifications
| Chemical Name or Material | N-(6-(3-((N,N-diethylsulfamoyl)amino)phenyl)-1H-indazol-3-yl)cyclopropanecarboxamide |
| CAS | 2247894-32-0 |
| Quantity | 25 mg |
| Molecular Formula | C21H25N5O3S |
| Formula Weight | 427.52 |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction