missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ SGC AAK1 1
GREENER_CHOICE

Product Code. 18798376
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
This item is not returnable. View return policy

Product Code. 18798376

Brand: Tocris Bioscience™ 6528/25

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

AAK1 and BMP2K inhibitor; activates Wnt signaling

SGC AAK1 1 is an adaptor-associated protein kinase 1 (AAK1) and BMP-2-inducible protein kinase (BMP2K) inhibitor (IC50 values are 233 nM and 1.48 μM for AAK1 and BMP2K, respectively). Inhibits AAK1-dependent phosphorylation of AP2M1. Activates Wnt signaling.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-(6-(3-((N,N-diethylsulfamoyl)amino)phenyl)-1H-indazol-3-yl)cyclopropanecarboxamide
CAS 2247894-32-0
Quantity 25 mg
Molecular Formula C21H25N5O3S
Formula Weight 427.52
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.